This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OVOL2 SIRNA Assays ›
Gene Symbol:
Ovol2
Gene Name:
ovo-like zinc finger 2
Gene Aliases:
Zfp339
Chromosome Location:
Chr.3: 138433990 - 138463690 on Build Rnor_6.0
UniGene ID:
Rn.104639
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s149944

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ovol2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106519.1 NP_001099989.1 4 689
XM_006235124.2 XP_006235186.1 4 1848

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
neural crest cell migration
neural fold formation
heart looping
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
heart development
negative regulation of cell proliferation
epidermis development
epidermal cell differentiation
dorsal/ventral pattern formation
negative regulation of epithelial to mesenchymal transition
regulation of keratinocyte proliferation
negative regulation of transcription by competitive promoter binding
negative regulation of keratinocyte differentiation
positive regulation of keratinocyte differentiation
negative regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
embryonic digestive tract morphogenesis
regulation of cell cycle
endocardium formation
heart trabecula formation
labyrinthine layer blood vessel development
negative regulation of stem cell proliferation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline