siRNA Details
| Gene Symbol: Zfp966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762405.1 | XP_008760627.1 | 3 | 224 | ||
| GenBank mRNA | AY539896.1 | - | 5 | 545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp966| Gene Symbol: Zfp966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762405.1 | XP_008760627.1 | 3 | 224 | ||
| GenBank mRNA | AY539896.1 | - | 5 | 545 | ||