siRNA Details
| Gene Symbol: Clec2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044233.2 | NP_001037698.2 | 5 | 577 | ||
| GenBank mRNA | BC161812.1 | - | 707 | |||
| DQ630549.1 | - | 5 | 437 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec2l| Gene Symbol: Clec2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044233.2 | NP_001037698.2 | 5 | 577 | ||
| GenBank mRNA | BC161812.1 | - | 707 | |||
| DQ630549.1 | - | 5 | 437 | |||