This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF84 SIRNA Assays ›
Gene Symbol:
ZNF84
Gene Name:
zinc finger protein 84
Gene Aliases:
HPF2
Chromosome Location:
Chr.12: 133037286 - 133063304 on Build GRCh38
UniGene ID:
Hs.445019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s15133

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF84
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127372.2 NP_001120844.1 5 2186
NM_001289971.1 NP_001276900.1 5 2177
NM_001289972.1 NP_001276901.1 5 2305
NM_003428.5 NP_003419.3 5 2170
XM_005266185.1 XP_005266242.1 5 2340
XM_005266186.1 XP_005266243.1 6 2076
XM_011534832.1 XP_011533134.1 5 2120
GenBank mRNA AK314232.1 - 5 1948
BC036656.1 - 5 2292 4828620
BC048350.1 - 4 1754 5555056
BC062552.1 - 5 1946 6536224
DN601581.1 - 1 711
M27878.1 - 5 2078
X60156.2 - 4 1916

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-l4n7g:80/100.66.72.160:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0