This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF135 SIRNA Assays ›
Gene Symbol:
ZNF135
Gene Name:
zinc finger protein 135
Gene Aliases:
ZNF61, ZNF78L1, pHZ-17, pT3
Chromosome Location:
Chr.19: 58059227 - 58069755 on Build GRCh38
UniGene ID:
Hs.85863
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s15171

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF135
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164530.1 NP_001158002.1 3, 4 305
NM_001289401.1 NP_001276330.1 4, 5 372
NM_001289402.1 NP_001276331.1 4, 5 735
NM_003436.3 NP_003427.3 5 403
NM_007134.1 NP_009065.1 4 341
XM_005259211.1 XP_005259268.1 3, 4 305
XM_006723362.3 XP_006723425.1 5 411
XM_006723363.3 XP_006723426.1 5 775
XM_017027240.1 XP_016882729.1 3, 4 728
XM_017027241.1 XP_016882730.1 5 730
XM_017027242.1 XP_016882731.1 3 226
GenBank mRNA AK094202.1 - 4, 5 694
AK098011.1 - 3, 4 305
AK295110.1 - 5 373
BC046434.1 - 4, 5 367 4824328
BX647775.1 - 4 309
DB117846.1 - 3, 4 305

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cytoskeleton organization
regulation of cell morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline