This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF135 SIRNA Assays ›
Gene Symbol:
ZNF135
Gene Name:
zinc finger protein 135
Gene Aliases:
ZNF61, ZNF78L1, pHZ-17, pT3
Chromosome Location:
Chr.19: 58059227 - 58069755 on Build GRCh38
UniGene ID:
Hs.85863
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s15172

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF135
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164529.1 NP_001158001.1 4 1501
NM_001289401.1 NP_001276330.1 5 1927
NM_001289402.1 NP_001276331.1 5 2290
NM_003436.3 NP_003427.3 5 1958
NM_007134.1 NP_009065.1 4 1896
XM_005259211.1 XP_005259268.1 4 1860
XM_006723362.3 XP_006723425.1 5 1966
XM_006723363.3 XP_006723426.1 5 2330
XM_017027240.1 XP_016882729.1 4 2283
XM_017027241.1 XP_016882730.1 5 2285
XM_017027242.1 XP_016882731.1 4 1725
GenBank mRNA AK094202.1 - 5 2249
AK295110.1 - 5 1928
AK297226.1 - 4 1691
AL833637.1 - 4 1459
BC046434.1 - 5 1922 4824328
BX647775.1 - 4 1864
U09413.1 - 1 1329

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cytoskeleton organization
regulation of cell morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline