siRNA Details
| Gene Symbol: Flrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106750.1 | NP_001100220.1 | 2 | 1553 | ||
| XM_006240416.2 | XP_006240478.1 | 1 | 1188 | |||
| XM_008764760.1 | XP_008762982.1 | 2 | 1248 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Flrt2| Gene Symbol: Flrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106750.1 | NP_001100220.1 | 2 | 1553 | ||
| XM_006240416.2 | XP_006240478.1 | 1 | 1188 | |||
| XM_008764760.1 | XP_008762982.1 | 2 | 1248 | |||