siRNA Details
| Gene Symbol: Rab21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004238.1 | NP_001004238.1 | 3, 4 | 535 | ||
| GenBank mRNA | BC079323.1 | - | 3, 4 | 535 | 7135453 | |
| FQ226410.1 | - | 2, 3 | 137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab21| Gene Symbol: Rab21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004238.1 | NP_001004238.1 | 3, 4 | 535 | ||
| GenBank mRNA | BC079323.1 | - | 3, 4 | 535 | 7135453 | |
| FQ226410.1 | - | 2, 3 | 137 | |||