siRNA Details
| Gene Symbol: Morf4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011999.1 | NP_001011999.1 | 10 | 806 | ||
| XM_008766445.1 | XP_008764667.1 | 11 | 889 | |||
| GenBank mRNA | BC078910.1 | - | 10 | 806 | 7131534 | |
| FQ213551.1 | - | 4 | 326 | |||

Morf4l1
Genomic Map