siRNA Details
| Gene Symbol: Gltscr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106888.1 | NP_001100358.1 | 5 | 1327 | ||
| XM_006244508.2 | XP_006244570.1 | 4 | 3065 | |||
| XM_008766846.1 | XP_008765068.1 | 6 | 1389 | |||
| XM_008766847.1 | XP_008765069.1 | 5 | 1353 | |||

Gltscr1l
Genomic Map