siRNA Details
| Gene Symbol: Polr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008330.1 | NP_001008331.1 | 3 | 294 | ||
| GenBank mRNA | BC086597.1 | - | 3 | 294 | 7312277 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr1c| Gene Symbol: Polr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008330.1 | NP_001008331.1 | 3 | 294 | ||
| GenBank mRNA | BC086597.1 | - | 3 | 294 | 7312277 | |