siRNA Details
| Gene Symbol: Txlng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037187.1 | NP_001032264.1 | 5 | 851 | ||
| XM_006256890.2 | XP_006256952.1 | 5 | 911 | |||
| XM_008773168.1 | XP_008771390.1 | 5 | 911 | |||
| GenBank mRNA | BC099110.1 | - | 5 | 851 | 7461546 | |

Txlng
Genomic Map