siRNA Details
| Gene Symbol: Smptb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182818.1 | NP_877970.1 | 1 | 1052 | ||
| GenBank mRNA | AY223520.1 | - | 1 | 1052 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smptb| Gene Symbol: Smptb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182818.1 | NP_877970.1 | 1 | 1052 | ||
| GenBank mRNA | AY223520.1 | - | 1 | 1052 | ||