siRNA Details
| Gene Symbol: Telo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_220236.7 | XP_220236.3 | 18 | 2394 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Telo2| Gene Symbol: Telo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_220236.7 | XP_220236.3 | 18 | 2394 | ||