siRNA Details
| Gene Symbol: Sgca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107039.1 | NP_001100509.1 | 10 | 1255 | ||
| XM_006247191.2 | XP_006247253.1 | 9 | 1079 | |||
| XM_006247192.1 | XP_006247254.1 | 9 | 983 | |||
| GenBank mRNA | FQ212993.1 | - | 8 | 886 | ||
| FQ215532.1 | - | 9 | 1041 | |||

Sgca
Genomic Map