siRNA Details
| Gene Symbol: Osbpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107044.1 | NP_001100514.1 | 18 | 2116 | ||
| XM_006247299.2 | XP_006247361.1 | 19 | 2173 | |||
| XM_006247300.2 | XP_006247362.1 | 19 | 2113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl7| Gene Symbol: Osbpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107044.1 | NP_001100514.1 | 18 | 2116 | ||
| XM_006247299.2 | XP_006247361.1 | 19 | 2173 | |||
| XM_006247300.2 | XP_006247362.1 | 19 | 2113 | |||