siRNA Details
| Gene Symbol: Arhgap23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247437.1 | XP_006247499.1 | 5 | 1024 | ||
| XM_006247438.1 | XP_006247500.1 | 5 | 1024 | |||
| XM_008768198.1 | XP_008766420.1 | 4 | 1484 | |||
| XM_008768199.1 | XP_008766421.1 | 4 | 1484 | |||
| XM_008768200.1 | XP_008766422.1 | 3 | 2479 | |||
| XM_220918.8 | XP_220918.6 | 7 | 1328 | |||

Arhgap23
Genomic Map