siRNA Details
| Gene Symbol: Engase | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107070.1 | NP_001100540.1 | 2, 3 | 346 | ||
| XM_006247803.2 | XP_006247865.1 | 2, 3 | 333 | |||
| XM_006247804.2 | XP_006247866.1 | 2, 3 | 158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Engase| Gene Symbol: Engase | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107070.1 | NP_001100540.1 | 2, 3 | 346 | ||
| XM_006247803.2 | XP_006247865.1 | 2, 3 | 333 | |||
| XM_006247804.2 | XP_006247866.1 | 2, 3 | 158 | |||