siRNA Details
| Gene Symbol: Cbx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034078.1 | NP_001029250.1 | 2 | 96 | ||
| GenBank mRNA | BC099206.1 | - | 2 | 96 | 7384263 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbx8| Gene Symbol: Cbx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034078.1 | NP_001029250.1 | 2 | 96 | ||
| GenBank mRNA | BC099206.1 | - | 2 | 96 | 7384263 | |