siRNA Details
| Gene Symbol: Klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181473.2 | NP_852138.2 | 5, 6 | 1569 | ||
| GenBank mRNA | AB101615.1 | - | 4, 5 | 1421 | ||
| BC092204.1 | - | 5, 6 | 1569 | 7312023 | ||
| FQ219171.1 | - | 6, 7 | 1652 | |||

Klhl24
Genomic Map