siRNA Details
| Gene Symbol: Bcl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107084.1 | NP_001100554.1 | 5 | 1536 | ||
| XM_008768799.1 | XP_008767021.1 | 4 | 2409 | |||
| GenBank mRNA | BC166425.1 | - | 5 | 1436 | 7097735 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl6| Gene Symbol: Bcl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107084.1 | NP_001100554.1 | 5 | 1536 | ||
| XM_008768799.1 | XP_008767021.1 | 4 | 2409 | |||
| GenBank mRNA | BC166425.1 | - | 5 | 1436 | 7097735 | |