siRNA Details
| Gene Symbol: Cpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107085.2 | NP_001100555.1 | 2 | 877 | ||
| XM_006248485.2 | XP_006248547.1 | 3 | 934 | |||
| GenBank mRNA | BC160859.1 | - | 1 | 813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpn2| Gene Symbol: Cpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107085.2 | NP_001100555.1 | 2 | 877 | ||
| XM_006248485.2 | XP_006248547.1 | 3 | 934 | |||
| GenBank mRNA | BC160859.1 | - | 1 | 813 | ||