This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CPN2 SIRNA Assays ›
Gene Symbol:
Cpn2
Gene Name:
carboxypeptidase N, polypeptide 2
Gene Aliases:
RGD1305170
Chromosome Location:
Chr.11: 74057374 - 74067486 on Build Rnor_6.0
UniGene ID:
Rn.126621
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s155594

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cpn2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107085.2 NP_001100555.1 2 655
XM_006248485.2 XP_006248547.1 3 712
GenBank mRNA BC160859.1 - 1 591

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline