siRNA Details
| Gene Symbol: Cep97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107096.1 | NP_001100566.1 | 4 | 483 | ||
| XM_006248250.1 | XP_006248312.1 | 2 | 158 | |||
| XM_008768642.1 | XP_008766864.1 | 4 | 490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep97| Gene Symbol: Cep97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107096.1 | NP_001100566.1 | 4 | 483 | ||
| XM_006248250.1 | XP_006248312.1 | 2 | 158 | |||
| XM_008768642.1 | XP_008766864.1 | 4 | 490 | |||