siRNA Details
| Gene Symbol: Daglb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107120.1 | NP_001100590.1 | 9 | 1160 | ||
| XM_008768961.1 | XP_008767183.1 | 9 | 1264 | |||
| GenBank mRNA | FQ212319.1 | - | 9 | 1174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Daglb| Gene Symbol: Daglb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107120.1 | NP_001100590.1 | 9 | 1160 | ||
| XM_008768961.1 | XP_008767183.1 | 9 | 1264 | |||
| GenBank mRNA | FQ212319.1 | - | 9 | 1174 | ||