siRNA Details
| Gene Symbol: RGD1566386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107127.2 | NP_001100597.2 | 4 | 657 | ||
| XM_008768973.1 | XP_008767195.1 | 4 | 661 | |||
| GenBank mRNA | BC167756.1 | - | 4 | 657 | 7099544 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1566386| Gene Symbol: RGD1566386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107127.2 | NP_001100597.2 | 4 | 657 | ||
| XM_008768973.1 | XP_008767195.1 | 4 | 661 | |||
| GenBank mRNA | BC167756.1 | - | 4 | 657 | 7099544 | |