siRNA Details
| Gene Symbol: LOC100910636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751148.3 | XP_003751196.1 | 10, 11 | 1282 | ||
| XM_006249080.2 | XP_006249142.1 | 10, 11 | 1280 | |||
| XM_008769105.1 | XP_008767327.1 | 10, 11 | 1273 | |||
| Gene Symbol: Zcwpw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249069.2 | XP_006249131.1 | 9, 10 | 1439 | ||

LOC100910636
Genomic Map