siRNA Details
| Gene Symbol: Sptbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013130.1 | NP_001013148.1 | 8, 9 | 1012 | ||
| XM_008770458.1 | XP_008768680.1 | 8, 9 | 2549 | |||
| XM_008770460.1 | XP_008768682.1 | 7, 8 | 2406 | |||
| XM_008770461.1 | XP_008768683.1 | 8, 9 | 1029 | |||
| XM_008770462.1 | XP_008768684.1 | 7, 8 | 2406 | |||
| XM_008770463.1 | XP_008768685.1 | 8, 9 | 2549 | |||
| GenBank mRNA | AY238344.1 | - | 8, 9 | 1012 | ||

Sptbn1
Genomic Map