This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SAMD4A SIRNA Assays ›
Gene Symbol:
Samd4a
Gene Name:
sterile alpha motif domain containing 4A
Gene Aliases:
Samd4
Chromosome Location:
Chr.15: 23777000 - 23879067 on Build Rnor_6.0
UniGene ID:
Rn.224832
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s157020

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Samd4a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107254.1 NP_001100724.1 2 405
XM_008770515.1 XP_008768737.1 2 1243
XM_008770516.1 XP_008768738.1 2 1242
GenBank mRNA BC168894.1 - 2 387 7453870

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK134243.

Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
negative regulation of translation
positive regulation of translation
translation repressor activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-tf58v:80/100.66.78.228:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0