siRNA Details
| Gene Symbol: Amt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014004.1 | NP_001014026.1 | 2 | 388 | ||
| GenBank mRNA | BC083803.1 | - | 2 | 388 | 7109120 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amt| Gene Symbol: Amt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014004.1 | NP_001014026.1 | 2 | 388 | ||
| GenBank mRNA | BC083803.1 | - | 2 | 388 | 7109120 | |