siRNA Details
| Gene Symbol: Zbtb45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107478.1 | NP_001100948.1 | 3 | 2155 | ||
| XM_006228114.2 | XP_006228176.1 | 3 | 3416 | |||
| XM_006228115.2 | XP_006228177.1 | 3 | 3324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb45| Gene Symbol: Zbtb45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107478.1 | NP_001100948.1 | 3 | 2155 | ||
| XM_006228114.2 | XP_006228176.1 | 3 | 3416 | |||
| XM_006228115.2 | XP_006228177.1 | 3 | 3324 | |||