siRNA Details
| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107557.1 | NP_001101027.1 | 4 | 587 | ||
| XM_006230436.2 | XP_006230498.1 | 4 | 1133 | |||
| XM_008759889.1 | XP_008758111.1 | 4 | 1135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Edrf1| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107557.1 | NP_001101027.1 | 4 | 587 | ||
| XM_006230436.2 | XP_006230498.1 | 4 | 1133 | |||
| XM_008759889.1 | XP_008758111.1 | 4 | 1135 | |||