siRNA Details
| Gene Symbol: Frmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008348.1 | NP_001008349.1 | 3, 4 | 418 | ||
| XM_006230804.2 | XP_006230866.1 | 3, 4 | 442 | |||
| GenBank mRNA | BC085896.1 | - | 3, 4 | 418 | 7193163 | |
| FQ211651.1 | - | 3, 4 | 457 | |||

Frmd8
Genomic Map