siRNA Details
| Gene Symbol: Tspyl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012075.1 | NP_001012075.1 | 1 | 771 | ||
| GenBank mRNA | BC082023.1 | - | 1 | 771 | 7129838 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspyl4| Gene Symbol: Tspyl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012075.1 | NP_001012075.1 | 1 | 771 | ||
| GenBank mRNA | BC082023.1 | - | 1 | 771 | 7129838 | |