siRNA Details
| Gene Symbol: Fam198b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199105.1 | NP_954536.1 | 4 | 1607 | ||
| XM_006232514.2 | XP_006232576.1 | 5 | 1938 | |||
| GenBank mRNA | BC061747.1 | - | 4 | 1607 | 5599685 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam198b| Gene Symbol: Fam198b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199105.1 | NP_954536.1 | 4 | 1607 | ||
| XM_006232514.2 | XP_006232576.1 | 5 | 1938 | |||
| GenBank mRNA | BC061747.1 | - | 4 | 1607 | 5599685 | |