siRNA Details
| Gene Symbol: Golph3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007698.1 | NP_001007699.1 | 5 | 624 | ||
| GenBank mRNA | BC081987.1 | - | 5 | 624 | 7129067 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Golph3l| Gene Symbol: Golph3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007698.1 | NP_001007699.1 | 5 | 624 | ||
| GenBank mRNA | BC081987.1 | - | 5 | 624 | 7129067 | |