siRNA Details
| Gene Symbol: Rsbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191710.1 | NP_001178639.1 | 3 | 1757 | ||
| XM_006233082.2 | XP_006233144.1 | 3 | 1456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsbn1| Gene Symbol: Rsbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191710.1 | NP_001178639.1 | 3 | 1757 | ||
| XM_006233082.2 | XP_006233144.1 | 3 | 1456 | |||