siRNA Details
| Gene Symbol: Palmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025688.1 | NP_001020859.1 | 669 | |||
| GenBank mRNA | BC098755.1 | - | 669 | 7370232 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Palmd| Gene Symbol: Palmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025688.1 | NP_001020859.1 | 669 | |||
| GenBank mRNA | BC098755.1 | - | 669 | 7370232 | ||