siRNA Details
| Gene Symbol: Btbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107782.1 | NP_001101252.1 | 4 | 1349 | ||
| XM_006235099.2 | XP_006235161.1 | 5 | 1106 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd3| Gene Symbol: Btbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107782.1 | NP_001101252.1 | 4 | 1349 | ||
| XM_006235099.2 | XP_006235161.1 | 5 | 1106 | |||