siRNA Details
| Gene Symbol: Sel1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014049.1 | NP_001014071.1 | 9 | 987 | ||
| GenBank mRNA | BC083893.1 | - | 9 | 987 | 7132857 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sel1l2| Gene Symbol: Sel1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014049.1 | NP_001014071.1 | 9 | 987 | ||
| GenBank mRNA | BC083893.1 | - | 9 | 987 | 7132857 | |