This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PHF20 SIRNA Assays ›
Gene Symbol:
Phf20
Gene Name:
PHD finger protein 20
Gene Aliases:
RGD1305020
Chromosome Location:
Chr.3: 152273269 - 152379742 on Build Rnor_6.0
UniGene ID:
Rn.14290
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s161298

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Phf20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107795.1 NP_001101265.1 17 3058
XM_006235404.2 XP_006235466.1 18 3134
XM_008762342.1 XP_008760564.1 18 3145

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
DNA binding
zinc ion binding
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0