siRNA Details
| Gene Symbol: Stk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107800.1 | NP_001101270.1 | 6 | 652 | ||
| GenBank mRNA | FQ226333.1 | - | 2 | 231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk4| Gene Symbol: Stk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107800.1 | NP_001101270.1 | 6 | 652 | ||
| GenBank mRNA | FQ226333.1 | - | 2 | 231 | ||