siRNA Details
| Gene Symbol: Rexo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033884.1 | NP_001029056.1 | 5, 6 | 1030 | ||
| GenBank mRNA | BC161820.1 | - | 5, 6 | 995 | 9088929 | |
| CB611468.1 | - | 4, 5 | 515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rexo4| Gene Symbol: Rexo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033884.1 | NP_001029056.1 | 5, 6 | 1030 | ||
| GenBank mRNA | BC161820.1 | - | 5, 6 | 995 | 9088929 | |
| CB611468.1 | - | 4, 5 | 515 | |||