siRNA Details
| Gene Symbol: Scaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271170.1 | NP_001258099.1 | 12 | 3597 | ||
| GenBank mRNA | AY724482.1 | - | 3 | 2010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scaf11| Gene Symbol: Scaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271170.1 | NP_001258099.1 | 12 | 3597 | ||
| GenBank mRNA | AY724482.1 | - | 3 | 2010 | ||