This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZFP746 SIRNA Assays ›
Gene Symbol:
Zfp746
Gene Name:
zinc finger protein 746
Gene Aliases:
RGD1306209, Znf746
Chromosome Location:
Chr.4: 77723182 - 77747512 on Build Rnor_6.0
UniGene ID:
Rn.48855
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s161758

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp746
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100852.1 NP_001094322.1 7 1428
XM_006236419.2 XP_006236481.1 7 1867
XM_008762885.1 XP_008761107.1 6 3759
XM_008762886.1 XP_008761108.1 6 3756
XM_008762887.1 XP_008761109.1 7 1862
XM_008762888.1 XP_008761110.1 5 3633
GenBank mRNA BC087150.1 - 1 289

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of neuron death
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin protein ligase binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-l5nwv:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline