This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CD163 SIRNA Assays ›
Gene Symbol:
Cd163
Gene Name:
CD163 molecule
Gene Aliases:
-
Chromosome Location:
Chr.4: 156752064 - 156785467 on Build Rnor_6.0
UniGene ID:
Rn.203204
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s161953

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd163
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107887.1 NP_001101357.1 7, 8 1829
XM_006237352.2 XP_006237414.1 7, 8 1858

Back To Top

More Information


Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
scavenger receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-s62d2:80/100.66.75.204:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0