siRNA Details
| Gene Symbol: Gsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013166.1 | NP_001013184.2 | 6 | 1211 | ||
| GenBank mRNA | BC079000.1 | - | 6 | 1211 | 7112802 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsg1| Gene Symbol: Gsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013166.1 | NP_001013184.2 | 6 | 1211 | ||
| GenBank mRNA | BC079000.1 | - | 6 | 1211 | 7112802 | |