This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZFP618 SIRNA Assays ›
Gene Symbol:
Zfp618
Gene Name:
zinc finger protein 618
Gene Aliases:
Znf618
Chromosome Location:
Chr.5: 78806604 - 78972433 on Build Rnor_6.0
UniGene ID:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s162322

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp618
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763811.1 XP_008762033.1 16 2893
XM_008763812.1 XP_008762034.1 16 2890
XM_008763813.1 XP_008762035.1 16 2869
XM_008763814.1 XP_008762036.1 15 2848
XM_008763815.1 XP_008762037.1 15 2797
XM_008763816.1 XP_008762038.1 15 2773

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028326, AK173309, AK012894.

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Process(es)

nucleic acid binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-4spjb:80/100.66.76.244:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0