This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IFT74 SIRNA Assays ›
Gene Symbol:
Ift74
Gene Name:
intraflagellar transport 74
Gene Aliases:
Ccdc2
Chromosome Location:
Chr.5: 113592919 - 113682485 on Build Rnor_6.0
UniGene ID:
Rn.15267
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s162383

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ift74
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007001.1 NP_001007002.1 8 708
XM_006238398.2 XP_006238460.1 8 816
GenBank mRNA BC083611.1 - 8 708 7133514

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

keratinocyte development
Notch signaling pathway
epidermis development
positive regulation of cell adhesion mediated by integrin
intraciliary transport involved in cilium morphogenesis
intraciliary transport
cilium assembly
positive regulation of transcription from RNA polymerase II promoter
negative regulation of epithelial cell proliferation
cilium morphogenesis
chromatin binding
beta-tubulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-245sx:80/100.66.79.25:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0