siRNA Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191770.1 | NP_001178699.1 | 12, 13 | 2209 | ||
| XM_008764459.1 | XP_008762681.1 | 13, 14 | 2638 | |||
| XM_008764460.1 | XP_008762682.1 | 13, 14 | 2346 | |||
| XM_008764461.1 | XP_008762683.1 | 13, 14 | 2338 | |||
| XM_008764462.1 | XP_008762684.1 | 12, 13 | 2717 | |||
| XM_008764463.1 | XP_008762685.1 | 11, 12 | 2143 | |||

Plekhh2
Genomic Map